accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 198084-13-8

OHNN

CAS RN 198084-13-8

IUPAC Name: [4-(pyrimidin-5-yl)phenyl]methanol
Synonyms: [4-(pyrimidin-5-yl)phenyl]methanol
Molecular Weight (g/mol): 186.21
Molecular Formula: C11H10N2O
InChi Key: QPOIDCHBWLGFGU-UHFFFAOYSA-N
SMILES: OCC1=CC=C(C=C1)C1=CN=CN=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results