accessibility menu, dialog, popup

UserName

Viewing profile as user:
N(R)PHN(R)OPOOP(R)(S)OOOOOOOOOOHHOHONaNaNa

CAS RN 19817-92-6

IUPAC Name: trisodium ({[(2R,3S,4R,5R)-5-(2,4-dioxo-1,2,3,4-tetrahydropyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphonato}oxy)(hydrogen phosphonatooxy)phosphinate
Synonyms: uridine 5'-triphosphate sodium
Molecular Weight (g/mol): 550.09
Molecular Formula: C9H12N2Na3O15P3
InChi Key: MMJGIWFJVDOPJF-ZKGVITBDNA-K
SMILES: [Na+].[Na+].[Na+].O[C@H]1[C@@H](O)[C@@H](O[C@@H]1COP([O-])(=O)OP([O-])(=O)OP(O)([O-])=O)N1C=CC(=O)NC1=O
Quantity 
  • (3)
  • (3)
  • (1)
  • (3)
  • (2)
Physical Form 
  • (4)
  • (4)
Percent Purity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
Grade 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (9)
  • (3)
  • (3)
  • (3)
  • (1)
  • (3)
  • (2)
  • (4)
  • (4)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)