Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Uridine-5'-Triphosphate Trisodium salt, 90 to 95%, MP Biomedicals™

Catalog No. ICN19123091 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
1 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19123091 1 g
ICN19123080 100 mg
ICN19123083 250 mg
ICN19123090 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. ICN19123091 Supplier MP Biomedicals, Inc Supplier No. 0219123091
Add to Cart
Add to Cart

Chemical Identifiers

CAS 19817-92-6
Molecular Formula C9H12N2Na3O15P3
Molecular Weight (g/mol) 550.09
MDL Number MFCD11045990
InChI Key MMJGIWFJVDOPJF-ZKGVITBDNA-K
Synonym uridine-5'-triphosphate trisodium salt dihydrate, uridine 5'-triphosphate trisodium salt dihydrate, trisodium uridine 5'-triphosphate dihydrate, 550.0866 anhydrous, c9h12n2na3o15p3.xh2o, uridine 5'-tetrahydrogen triphosphate , sodium salt, hydrate 1:3:2, trisodium dihydrate utp trianion, sodium 5'-o-hydroxyphosphinato oxy phosphinato oxy phosphinato uridine hydrate 3:1:2, trisodium, 2r,3s,4r,5r-5-2,4-dioxopyrimidin-1-yl-3,4-dihydroxyoxolan-2-yl methoxy-oxidophosphoryl oxy-oxidophosphoryl hydrogen phosphate, dihydrate, uridine 5'-triphosphate, trisodium salt, hydrate
PubChem CID 131675549
IUPAC Name trisodium ({[(2R,3S,4R,5R)-5-(2,4-dioxo-1,2,3,4-tetrahydropyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphonato}oxy)(hydrogen phosphonatooxy)phosphinate
SMILES [Na+].[Na+].[Na+].O[C@H]1[C@@H](O)[C@@H](O[C@@H]1COP([O-])(=O)OP([O-])(=O)OP(O)([O-])=O)N1C=CC(=O)NC1=O

Specifications

Physical Form Solid
Quantity 1 g
Assay Percent Range 90 to 95%
Formula Weight 550.09
Appearance Powder
Chemical Name or Material Uridine-5'-Triphosphate Trisodium salt
EINECSNumber 243-347-5
Recommended Storage Store at 0°C.
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.