accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3HO

CAS RN 19819-98-8

IUPAC Name: 2-(2-methylphenyl)ethan-1-ol
Synonyms: benzeneethanol, 2-methyl-
Molecular Weight (g/mol): 136.19
Molecular Formula: C9H12O
InChi Key: RUGISKODRCWQNE-UHFFFAOYSA-N
SMILES: CC1=CC=CC=C1CCO
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results