Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(o-Tolyl)ethanol 98.0+%, TCI America™
SDP

Catalog No. M13755G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M13755G 5 g
M137525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M13755G Supplier TCI America Supplier No. M13755G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 19819-98-8
Molecular Formula C9H12O
Molecular Weight (g/mol) 136.194
MDL Number MFCD00044241
InChI Key RUGISKODRCWQNE-UHFFFAOYSA-N
Synonym 2-o-tolyl ethanol, 2-methylphenethyl alcohol, 2-2-methylphenyl ethanol, benzeneethanol, 2-methyl, o-methylphenethyl alcohol, phenethyl alcohol, o-methyl, 2-methylbenzeneethanol, toluene-ethanol, unii-vx8700jgjp, 2-2-methylphenyl ethan-1-ol
PubChem CID 88267
IUPAC Name 2-(2-methylphenyl)ethanol
SMILES CC1=CC=CC=C1CCO

Specifications

Color Yellow
Boiling Point 243°C
Quantity 5 g
Formula Weight 136.19
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-(o-Tolyl)ethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.