accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1982-49-6

IUPAC Name: 3-(2-methylcyclohexyl)-1-phenylurea
Synonyms: siduron
Molecular Weight (g/mol): 232.33
Molecular Formula: C14H20N2O
InChi Key: JXVIIQLNUPXOII-UHFFFAOYNA-N
SMILES: CC1CCCCC1NC(=O)NC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results