accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1983-10-4

IUPAC Name: tributyltin fluoride
Synonyms: tributyltin fluoride
Molecular Weight (g/mol): 311.07
Molecular Formula: C12H29FSn
InChi Key: STUMXSORVXBBJR-UHFFFAOYSA-N
SMILES: F.CCCC[SnH](CCCC)CCCC
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)