accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 19833-78-4

IUPAC Name: diethylazanium iodide
Synonyms: diethylazanium iodide
Molecular Weight (g/mol): 201.05
Molecular Formula: C4H12IN
InChi Key: YYMLRIWBISZOMT-UHFFFAOYSA-N
SMILES: [I-].CC[NH2+]CC
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results