Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethylamine Hydroiodide 97.0+%, TCI America™
SDP

Catalog No. D46435G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D46435G 5 g
D46431G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D46435G Supplier TCI America Supplier No. D46435G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 19833-78-4
Molecular Formula C4H12IN
Molecular Weight (g/mol) 201.05
MDL Number MFCD28384142
InChI Key YYMLRIWBISZOMT-UHFFFAOYSA-N
Synonym Diethylammonium Iodide
PubChem CID 88320434
IUPAC Name diethylazanium iodide
SMILES [I-].CC[NH2+]CC

Specifications

Melting Point 175°C
Color White
Quantity 5 g
Formula Weight 201.05
Percent Purity ≥97.0% (N,T)
Physical Form Crystalline Powder
Chemical Name or Material Diethylamine Hydroiodide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.