accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3ONHSOONHI

CAS RN 19837-94-6

IUPAC Name: N-{4-[(4-iodophenyl)sulfamoyl]phenyl}acetamide
Synonyms: N-{4-[(4-iodophenyl)sulfamoyl]phenyl}acetamide
Molecular Weight (g/mol): 416.23
Molecular Formula: C14H13IN2O3S
InChi Key: VCYWXXUBOZNHBJ-UHFFFAOYSA-N
SMILES: CC(=O)NC1=CC=C(C=C1)S(=O)(=O)NC1=CC=C(I)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results