accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONONC

CAS RN 1984-23-2

IUPAC Name: 1-isocyano-4-nitrobenzene
Synonyms: 1-isocyano-4-nitrobenzene
Molecular Weight (g/mol): 148.12
Molecular Formula: C7H4N2O2
InChi Key: WQFPYEBGLBCQOY-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(C=C1)[N+]#[C-]
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results