Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Nitrophenyl Isocyanide 98.0+%, TCI America™
SDP

Catalog No. N09821G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N09821G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N09821G Supplier TCI America Supplier No. N09821G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1984-23-2
Molecular Formula C7H4N2O2
Molecular Weight (g/mol) 148.12
MDL Number MFCD01937394
InChI Key WQFPYEBGLBCQOY-UHFFFAOYSA-N
Synonym 1-Isocyano-4-nitrobenzene
PubChem CID 74802
IUPAC Name 1-isocyano-4-nitrobenzene
SMILES [O-][N+](=O)C1=CC=C(C=C1)[N+]#[C-]

Specifications

Melting Point 115°C
Color Yellow
Quantity 1 g
Formula Weight 148.12
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Nitrophenyl Isocyanide
RTECSNumber DA3768224
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.