accessibility menu, dialog, popup

UserName

Viewing profile as user:
NHNH

CAS RN 1984-49-2

IUPAC Name: 9H,9'H-3,3'-bicarbazole
Synonyms: 9H,9'H-3,3'-bicarbazole
Molecular Weight (g/mol): 332.41
Molecular Formula: C24H16N2
InChi Key: PUMOFXXLEABBTC-UHFFFAOYSA-N
SMILES: N1C2=C(C=CC=C2)C2=C1C=CC(=C2)C1=CC2=C(NC3=C2C=CC=C3)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results