Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,3'-Bicarbazole 98.0+%, TCI America™
SDP

Catalog No. B4646200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B4646200MG 200 mg
B46461G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B4646200MG Supplier TCI America Supplier No. B4646200MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1984-49-2
Molecular Formula C24H16N2
Molecular Weight (g/mol) 332.406
MDL Number MFCD11046470
InChI Key PUMOFXXLEABBTC-UHFFFAOYSA-N
PubChem CID 12462601
IUPAC Name 3-(9H-carbazol-3-yl)-9H-carbazole
SMILES C1=CC=C2C(=C1)C3=C(N2)C=CC(=C3)C4=CC5=C(C=C4)NC6=CC=CC=C65

Specifications

Quantity 200 mg
Formula Weight 332.41
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 3,3′-Bicarbazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.