accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 19847-10-0

IUPAC Name: pyrazine-2-carbonyl chloride
Synonyms: pyrazine-2-carbonyl chloride
Molecular Weight (g/mol): 142.54
Molecular Formula: C5H3ClN2O
InChi Key: TXJKATOSKLUITR-UHFFFAOYSA-N
SMILES: ClC(=O)C1=CN=CC=N1
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)