Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Pyrazinecarbonyl chloride, Technical Grade, Thermo Scientific™

Catalog No. CC03402DE
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC03402DE 5 g Amber Glass Bottle
CC03402DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC03402DE Supplier Thermo Scientific Chemicals Supplier No. CC03402DE
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 19847-10-0
Molecular Formula C5H3ClN2O
Molecular Weight (g/mol) 142.542
InChI Key TXJKATOSKLUITR-UHFFFAOYSA-N
Synonym 2-pyrazinecarbonyl chloride, pyrazinecarbonyl chloride, pyrazine-2-carboxylic acid chloride, 2-pyrazinecarbonylchloride, pyrazine 2-carbonyl chloride, 2-pyrazine carbonyl chloride, pyrazinoyl chloride, acmc-20ann0, 2-pyrazine carboxylic acid chloride, pyrazine-2-carbonyl chloride, technical 1g
PubChem CID 88279
IUPAC Name pyrazine-2-carbonyl chloride
SMILES C1=CN=C(C=N1)C(=O)Cl

Specifications

Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 142.54
Percent Purity Tech.
Grade Technical
Chemical Name or Material 2-Pyrazinecarbonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.