accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 198470-07-4

OHOOOHN(R)CH3H3CCH3HN

CAS RN 198470-07-4

IUPAC Name: (2R)-2-{[(tert-butoxy)carbonyl]amino}-3-cyclohexylpropanoic acid; N-cyclohexylcyclohexanamine
Synonyms: (2R)-2-[(tert-butoxycarbonyl)amino]-3-cyclohexylpropanoic acid; dicha
Molecular Weight (g/mol): 452.68
Molecular Formula: C26H48N2O4
InChi Key: GSGRQFGFPZQUQS-RFVHGSKJSA-N
SMILES: C1CCC(CC1)NC1CCCCC1.CC(C)(C)OC(=O)N[C@H](CC1CCCCC1)C(O)=O
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)