accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 198470-08-5

OHOOONH(R)CH3CH3H3CHN

CAS RN 198470-08-5

IUPAC Name: (2R)-2-{[(tert-butoxy)carbonyl]amino}-2-cyclohexylacetic acid; N-cyclohexylcyclohexanamine
Synonyms: (R)-[(tert-butoxycarbonyl)amino](cyclohexyl)acetic acid; dicha
Molecular Weight (g/mol): 438.65
Molecular Formula: C25H46N2O4
InChi Key: LHAPUSGSSYBOQR-HNCPQSOCSA-N
SMILES: C1CCC(CC1)NC1CCCCC1.CC(C)(C)OC(=O)N[C@H](C1CCCCC1)C(O)=O
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)