accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CH3CCH3OONH(S)FFOHO

CAS RN 198474-90-7

IUPAC Name: (2S)-2-{[(tert-butoxy)carbonyl]amino}-3-(3,4-difluorophenyl)propanoic acid
Synonyms: (2S)-2-[(tert-butoxycarbonyl)amino]-3-(3,4-difluorophenyl)propanoic acid
Molecular Weight (g/mol): 301.29
Molecular Formula: C14H17F2NO4
InChi Key: CYAOPVHXASZUDE-NSHDSACASA-N
SMILES: CC(C)(C)OC(=O)N[C@@H](CC1=CC=C(F)C(F)=C1)C(O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results