Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(tert-Butoxycarbonyl)-3,4-difluoro-L-phenylalanine 95.0+%, TCI America™
SDP

Catalog No. B36271G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B36271G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B36271G Supplier TCI America Supplier No. B36271G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 198474-90-7
Molecular Formula C14H17F2NO4
Molecular Weight (g/mol) 301.29
MDL Number MFCD00672518
InChI Key CYAOPVHXASZUDE-NSHDSACASA-N
Synonym boc-l-3,4-difluorophenylalanine, boc-phe 3,4-dif-oh, boc-3,4-difluoro-l-phenylalanine, boc-l-3,4-difluorophe, boc-phe 3,4-f2-oh, boc-3,4-difluorophenylalanine, n-boc-3,4-difluoro-l-phenylalanine, 2s-2-tert-butoxycarbonyl amino-3-3,4-difluorophenyl propanoic acid, boc-l-phe 3,4-f2-oh, 2s-2-tert-butoxycarbonyl amino-3-3,4-difluorophenyl propionic acid
PubChem CID 7020829
IUPAC Name (2S)-3-(3,4-difluorophenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
SMILES CC(C)(C)OC(=O)NC(CC1=CC(=C(C=C1)F)F)C(=O)O

Specifications

Melting Point 83°C
Color White-Yellow
Quantity 1 g
Formula Weight 301.29
Percent Purity ≥95.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-(tert-Butoxycarbonyl)-3,4-difluoro-L-phenylalanine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.