CAS RN 198545-46-9
IUPAC Name:
(2R)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-(2-fluorophenyl)propanoic acid
Synonyms:
(2R)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-3-(2-fluorophenyl)propanoic acid
Molecular Weight (g/mol):
405.43
Molecular Formula:
C24H20FNO4
InChi Key:
ARHOAMSIDCQWEW-JOCHJYFZSA-N
SMILES:
OC(=O)[C@@H](CC1=CC=CC=C1F)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
1
–
1
of
1
results
1
Advanced Chem Tech Fmoc-D-Phe 2F , Advanced ChemTech
CAS: 198545-46-9 Molecular Formula: C24H20FNO4 Molecular Weight (g/mol): 405.43 MDL Number: MFCD00672553 InChI Key: ARHOAMSIDCQWEW-JOCHJYFZSA-N IUPAC Name: (2R)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-(2-fluorophenyl)propanoic acid SMILES: OC(=O)[C@@H](CC1=CC=CC=C1F)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12