accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 198545-98-1

OOOHONHCH3(E)

CAS RN 198545-98-1

IUPAC Name: (2E)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)but-2-enoic acid
Synonyms: (2E)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}but-2-enoic acid
Molecular Weight (g/mol): 323.35
Molecular Formula: C19H17NO4
InChi Key: MCHXCZMMTWJODE-LAZPYJJCSA-N
SMILES: C\C=C(\NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2)C(O)=O
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)