CAS RN 198546-17-7
CAS RN 198546-17-7
IUPAC Name:
(2E)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-phenylprop-2-enoic acid
Synonyms:
(2E)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-3-phenylprop-2-enoic acid
Molecular Weight (g/mol):
385.42
Molecular Formula:
C24H19NO4
InChi Key:
QYKJHRNYJRVXQS-HYARGMPZSA-N
SMILES:
OC(=O)C(\NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2)=C/C1=CC=CC=C1
1
–
1
of
1
results
1
Advanced Chem Tech Fmoc-a-B-dehydro Phe, Advanced ChemTech
CAS: 198546-17-7 Molecular Formula: C24H19NO4 Molecular Weight (g/mol): 385.42 MDL Number: MFCD00235829 InChI Key: QYKJHRNYJRVXQS-HYARGMPZSA-N IUPAC Name: (2E)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-phenylprop-2-enoic acid SMILES: OC(=O)C(\NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2)=C/C1=CC=CC=C1