accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 198546-38-2

OOOHONHCH3H3C

CAS RN 198546-38-2

IUPAC Name: 2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-methylbut-2-enoic acid
Synonyms: 2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-3-methylbut-2-enoic acid
Molecular Weight (g/mol): 337.38
Molecular Formula: C20H19NO4
InChi Key: IYVIIHAWJFLOAR-UHFFFAOYSA-N
SMILES: CC(C)=C(NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2)C(O)=O
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)