CAS RN 198560-10-0
IUPAC Name:
(2S)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-4-(4-hydroxyphenyl)butanoic acid
Synonyms:
(2S)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-4-(4-hydroxyphenyl)butanoic acid
Molecular Weight (g/mol):
417.46
Molecular Formula:
C25H23NO5
InChi Key:
FZGWIJTZHIPMMG-QHCPKHFHSA-N
SMILES:
OC(=O)[C@H](CCC1=CC=C(O)C=C1)NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2
1
–
1
of
1
results
1
Fmoc-Htyr-OH Homotyrosine , Advanced ChemTech
CAS: 198560-10-0 Molecular Formula: C25H23NO5 Molecular Weight (g/mol): 417.46 MDL Number: MFCD00235904 InChI Key: FZGWIJTZHIPMMG-QHCPKHFHSA-N IUPAC Name: (2S)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-4-(4-hydroxyphenyl)butanoic acid SMILES: OC(=O)[C@H](CCC1=CC=C(O)C=C1)NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2