accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 198560-38-2

OOOHON

CAS RN 198560-38-2

IUPAC Name: 1-{[(9H-fluoren-9-yl)methoxy]carbonyl}-2,3-dihydro-1H-indole-2-carboxylic acid
Synonyms: 1-[(9H-fluoren-9-ylmethoxy)carbonyl]-2,3-dihydroindole-2-carboxylic acid
Molecular Weight (g/mol): 385.42
Molecular Formula: C24H19NO4
InChi Key: FWYAYXBFHLDIRA-UHFFFAOYSA-N
SMILES: OC(=O)C1CC2=C(C=CC=C2)N1C(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)