accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 198561-38-5

OOHOOONHNH(S)CH3CH3H3C

CAS RN 198561-38-5

IUPAC Name: (9H-fluoren-9-yl)methyl N-[(2S)-6-{[(tert-butoxy)carbonyl]amino}-1-hydroxyhexan-2-yl]carbamate
Synonyms: 9H-fluoren-9-ylmethyl N-[(2S)-6-[(tert-butoxycarbonyl)amino]-1-hydroxyhexan-2-yl]carbamate
Molecular Weight (g/mol): 454.57
Molecular Formula: C26H34N2O5
InChi Key: CQPCXWDXTZJRSX-SFHVURJKSA-N
SMILES: CC(C)(C)OC(=O)NCCCC[C@@H](CO)NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)