accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CH3CCH3OONHSOONOO

CAS RN 198572-71-3

IUPAC Name: tert-butyl N-(2-nitrobenzenesulfonyl)carbamate
Synonyms: tert-butyl N-(2-nitrobenzenesulfonyl)carbamate
Molecular Weight (g/mol): 302.3
Molecular Formula: C11H14N2O6S
InChi Key: WAYBGHIVTXTUCZ-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)NS(=O)(=O)C1=CC=CC=C1[N+]([O-])=O
Percent Purity 
  • (3)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results