Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(tert-Butoxycarbonyl)-2-nitrobenzenesulfonamide 98.0+%, TCI America™
SDP

Catalog No. B23035G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B23035G 5 g
B23031G 1 g
B230325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B23035G Supplier TCI America Supplier No. B23035G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 198572-71-3
Molecular Formula C11H14N2O6S
Molecular Weight (g/mol) 302.30
MDL Number MFCD06796217
InChI Key WAYBGHIVTXTUCZ-UHFFFAOYSA-N
Synonym N-Boc-2-nitrobenzenesulfonamide
PubChem CID 11141172
IUPAC Name tert-butyl N-(2-nitrobenzenesulfonyl)carbamate
SMILES CC(C)(C)OC(=O)NS(=O)(=O)C1=CC=CC=C1[N+]([O-])=O

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 302.30
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-(tert-Butoxycarbonyl)-2-nitrobenzenesulfonamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.