CAS RN 19870-46-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
phyproof™ 1,5-Dicaffeoylquinic acid, ≥98% (HPLC), MilliporeSigma™ Supelco™
In literature, CAS Numbers 19870-46-3 and 30964-13-7 are each described as 1,3-dicaffeoylquinic acid, 1,5-dicaffeoylquinic acid, and cynarin. The IUPAC synonym for this product is provided for clarification.