Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

phyproof™ 1,5-Dicaffeoylquinic acid, ≥98% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111002263 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-100-2263 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-100-2263 Supplier MilliporeSigma Supelco Supplier No. PHL8222110MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

In literature, CAS Numbers 19870-46-3 and 30964-13-7 are each described as 1,3-dicaffeoylquinic acid, 1,5-dicaffeoylquinic acid, and cynarin. The IUPAC synonym for this product is provided for clarification.

TRUSTED_SUSTAINABILITY

Specifications

CAS 19870-46-3
Product Line phyproof
Linear Formula C25H24O12
MDL Number MFCD13196505
Synonym (1 S,3 R,4 R,5 R)-1,3-Bis[3-(3,4-dihydroxyphenyl)acryloyloxy]-4,5-dihydroxycyclohexanecarboxylic acid; 1,3-Dicaffeoylquinic acid
Type Reference Substance
Quantity 10 mg
Formula Weight 516.45
Percent Purity ≥98% (HPLC)
Grade Primary Reference Standard

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.