accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOOHNOOH3C

CAS RN 19872-91-4

IUPAC Name: 2,6-dimethyl 4-oxo-1,4-dihydropyridine-2,6-dicarboxylate
Synonyms: 2,6-dimethyl 4-oxo-1H-pyridine-2,6-dicarboxylate
Molecular Weight (g/mol): 211.17
Molecular Formula: C9H9NO5
InChi Key: FERASHUCDLDGHK-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC(=O)C=C(N1)C(=O)OC
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results