CAS RN 19881-70-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ Windorphen
A cell-permeable, selective inhibitor p300 histone acetytransferase (IC50 = 4.2μM)