Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Windorphen

Catalog No. 5091640001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-916-40001 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-916-40001 Supplier MilliporeSigma™ Supplier No. 5091640001
Add to Cart
Add to Cart

A cell-permeable, selective inhibitor p300 histone acetytransferase (IC50 = 4.2μM)

The racemic mixture of a cell-permeable (4-methoxyphenyl)chloroacrylaldehyde compound whose ( Z )-isomer (isomer content 12-15%) acts as a p300-selective histone acetytransferase (HAT) inhibitor (IC50 = 4.2μM using racemic mixture with 13% Z-isomer) and selectively disrupts β-catenin association with p300, but not CBP or LEF1/TCF4, exhibiting much reduced potency against KAT5, CBP, MYST4, MYST2 (IC50 ≥38μM) and little or no inhibitory efficacy toward GCN5 and PCAF HAT activity (IC50 >100μM). Shown to selectively prevents human and mouse β-catenin-, zebrafish β-catenin-1-, but not zebrafish β-catenin-2-, mediated transcription activity without affecting β-catenin activation or nuclear translocation. Induces apoptosis in Wnt signaling-dependent cancer cultures (IC50 15 to 22μM; 72 h) in vitro without affecting the viability of Wnt-independent H460 (up to 200μM and 72 h) and selectively abrogates Wnt signaling in ventral and lateral regions, but not within dorsal organizer, in 5.3 hpf epiboly stage zebrafish embryos in vivo . A great complement to the selective β-catenin-CBP interaction blocker ICG-001 (Cat. no. 504712). ( E )-isomer is available separately (Cat. no.509166) as negative control.
TRUSTED_SUSTAINABILITY

Specifications

Color Off-white
Description 98% by HPLC
Target Proteins p300 HAT
CAS 19881-70-0
Molecular Formula C17H15ClO3
Solubility DMSO
Content And Storage Protect from light
Form Powder
Quantity 25 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.