accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 19900-72-2

IUPAC Name: 4-[(4-amino-3-ethyl-5-methylphenyl)methyl]-2-ethyl-6-methylaniline
Synonyms: 4-[(4-amino-3-ethyl-5-methylphenyl)methyl]-2-ethyl-6-methylaniline
Molecular Weight (g/mol): 282.43
Molecular Formula: C19H26N2
InChi Key: QJENIOQDYXRGLF-UHFFFAOYSA-N
SMILES: CCC1=CC(CC2=CC(C)=C(N)C(CC)=C2)=CC(C)=C1N
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results