Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4'-Methylenebis(2-ethyl-6-methylaniline) 98.0+%, TCI America™
SDP

Catalog No. M123825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M123825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M123825G Supplier TCI America Supplier No. M123825G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 19900-72-2
Molecular Formula C19H26N2
Molecular Weight (g/mol) 282.431
MDL Number MFCD00191227
InChI Key QJENIOQDYXRGLF-UHFFFAOYSA-N
Synonym Bis(4-amino-3-ethyl-5-methylphenyl)methane, 4,4′C-Diamino-3,3′C-diethyl-5,5′C-dimethyldiphenylmethane
PubChem CID 155611
IUPAC Name 4-[(4-amino-3-ethyl-5-methylphenyl)methyl]-2-ethyl-6-methylaniline
SMILES CCC1=CC(=CC(=C1N)C)CC2=CC(=C(C(=C2)CC)N)C

Specifications

Melting Point 85°C
Quantity 25 g
Formula Weight 282.43
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Methylenebis(2-ethyl-6-methylaniline)
EINECSNumber (4)-1651
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.