accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOH3COCH3OH3COHOH

CAS RN 19920-00-4

IUPAC Name: tetrakis(4-methoxyphenyl)ethane-1,2-diol
Synonyms: tetrakis(4-methoxyphenyl)ethane-1,2-diol
Molecular Weight (g/mol): 486.56
Molecular Formula: C30H30O6
InChi Key: GJSYWZLTIIFQTD-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1)C(O)(C1=CC=C(OC)C=C1)C(O)(C1=CC=C(OC)C=C1)C1=CC=C(OC)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results