Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1,2,2-Tetrakis(4-methoxyphenyl)-1,2-ethanediol 96.0+%, TCI America™
SDP

Catalog No. T16511G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T16511G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T16511G Supplier TCI America Supplier No. T16511G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 19920-00-4
Molecular Formula C30H30O6
Molecular Weight (g/mol) 486.564
MDL Number MFCD00185172
InChI Key GJSYWZLTIIFQTD-UHFFFAOYSA-N
Synonym 1,1,2,2-Tetra-p-anisyl-1,2-ethanediol
PubChem CID 3456938
IUPAC Name 1,1,2,2-tetrakis(4-methoxyphenyl)ethane-1,2-diol
SMILES COC1=CC=C(C=C1)C(C2=CC=C(C=C2)OC)(C(C3=CC=C(C=C3)OC)(C4=CC=C(C=C4)OC)O)O

Specifications

Melting Point 170°C
Color White
Quantity 1 g
Formula Weight 486.56
Percent Purity ≥96.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 1,1,2,2-Tetrakis(4-methoxyphenyl)-1,2-ethanediol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.