accessibility menu, dialog, popup

UserName

Viewing profile as user:
FFFFOO

CAS RN 19932-26-4

IUPAC Name: 2-[(2,2,3,3-tetrafluoropropoxy)methyl]oxirane
Synonyms: 2-[(2,2,3,3-tetrafluoropropoxy)methyl]oxirane
Molecular Weight (g/mol): 188.12
Molecular Formula: C6H8F4O2
InChi Key: DATKALAKXGFGPI-UHFFFAOYNA-N
SMILES: FC(F)C(F)(F)COCC1CO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results