Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Glycidyl 2,2,3,3-tetrafluoropropyl ether

Catalog No. 47415025ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
47-415-025ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 47-415-025ML Supplier Sigma Aldrich Supplier No. 47415025ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Glycidyl 2,2,3,3-tetrafluoropropyl ether

Specifications

CAS 19932-26-4
Density 1.327 g/mL (at 25°C)
Boiling Point 50°C (4 mmHg)
Molecular Formula C6H8F4O2
Refractive Index n20/D 1.366 (literature)
Linear Formula C6H8F4O2
Quantity 25 mL
MDL Number MFCD00054682
Molecular Weight (g/mol) 188.12
Percent Purity 97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.