accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 199328-31-9

IUPAC Name: 2-oxo-2,3-dihydro-1H-indole-5-sulfonyl chloride
Synonyms: 2-oxo-1,3-dihydroindole-5-sulfonyl chloride
Molecular Weight (g/mol): 231.65
Molecular Formula: C8H6ClNO3S
InChi Key: FYBNVKMJOOYPGI-UHFFFAOYSA-N
SMILES: ClS(=O)(=O)C1=CC2=C(NC(=O)C2)C=C1
Grade 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)