Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Oxoindoline-5-sulfonyl chloride, 97%, Thermo Scientific™

Catalog No. CC53303EA
Change view
Click to view available options
Quantity:
250 mg
1 g
10 g
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC53303EA 10 g Amber Glass Bottle
CC53303CB 250 mg Amber Glass Bottle
CC53303DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC53303EA Supplier Thermo Scientific Chemicals Supplier No. CC53303EA
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 199328-31-9
Molecular Formula C8H6ClNO3S
Molecular Weight (g/mol) 231.65
MDL Number MFCD06655924
InChI Key FYBNVKMJOOYPGI-UHFFFAOYSA-N
Synonym 2-oxoindoline-5-sulfonyl chloride, 2-oxo-2,3-dihydro-1h-indole-5-sulfonyl chloride, 2-oxoindoline-5-sulfonylchloride, 2-oxoindoline-5-sulphonyl chloride, 2,3-dihydro-2-oxo-1h-indole-5-sulphonyl chloride, 1h-indole-5-sulfonyl chloride, 2,3-dihydro-2-oxo, 5-chlorosulfonyl indolin-2-one, 5-chlorosulphonyloxindole, 5-chlorosulfonyl oxindole, 5-chlorosufonyl-2-oxindole
PubChem CID 4962506
SMILES ClS(=O)(=O)C1=CC2=C(NC(=O)C2)C=C1

Specifications

CAS Min % 80.0
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10 g
IUPAC Name 2-oxo-1,3-dihydroindole-5-sulfonyl chloride
Formula Weight 231.66
Percent Purity Tech.
Grade Technical
Chemical Name or Material 2-Oxoindoline-5-sulfonyl chloride
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Contact with water liberates toxic gas.
Reacts violently with water.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.