accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2CO

CAS RN 19955-99-8

IUPAC Name: 3-ethenylbenzaldehyde
Synonyms: 3-ethenylbenzaldehyde
Molecular Weight (g/mol): 132.16
Molecular Formula: C9H8O
InChi Key: CATOVPRCMWIZLR-UHFFFAOYSA-N
SMILES: C=CC1=CC(C=O)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results