accessibility menu, dialog, popup

UserName

Viewing profile as user:
FSF

CAS RN 1996-44-7

IUPAC Name: (2,4-difluorophenyl)sulfanide
Synonyms: (2,4-difluorophenyl)sulfanide
Molecular Weight (g/mol): 145.15
Molecular Formula: C6H3F2S
InChi Key: BICHBFCGCJNCAT-UHFFFAOYSA-M
SMILES: FC1=CC=C([S-])C(F)=C1
Boiling Point 
  • (2)
  • (1)
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)