Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Difluorobenzenethiol 97.0+%, TCI America™
SDP

Catalog No. D33425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D33425G 5 g
D334225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D33425G Supplier TCI America Supplier No. D33425G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1996-44-7
Molecular Formula C6H4F2S
Molecular Weight (g/mol) 146.155
InChI Key BICHBFCGCJNCAT-UHFFFAOYSA-N
Synonym 2,4-difluorothiophenol, 2,4-difluorobenzene-1-thiol, 2,4-difluoro thiophenol, benzenethiol, 2,4-difluoro, 2,4-difluorothiophenol technical, 2,4-difluorobenzenet, pubchem2875, acmc-1bpsf, 2,4-difluoro-benzenethiol
PubChem CID 2737073
IUPAC Name 2,4-difluorobenzenethiol
SMILES C1=CC(=C(C=C1F)F)S

Specifications

Color Yellow
Boiling Point 59°C
Quantity 5 g
UN Number 3336
Formula Weight 146.15
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,4-Difluorobenzenethiol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.