accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3ONHNOHNNHN

CAS RN 19962-37-9

IUPAC Name: N-(6-oxo-6,7-dihydro-3H-purin-2-yl)acetamide
Synonyms: N-(6-oxo-3,7-dihydropurin-2-yl)acetamide
Molecular Weight (g/mol): 193.17
Molecular Formula: C7H7N5O2
InChi Key: MXSMRDDXWJSGMC-UHFFFAOYSA-N
SMILES: CC(=O)NC1=NC(=O)C2=C(N1)N=CN2
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)