CAS RN 199682-73-0
CAS RN 199682-73-0
IUPAC Name:
5-(4-methoxyphenyl)-1H-pyrazole-4-carbaldehyde
Synonyms:
3-(4-methoxyphenyl)-2H-pyrazole-4-carbaldehyde
Molecular Weight (g/mol):
202.21
Molecular Formula:
C11H10N2O2
InChi Key:
QSGGFCPKXTULQQ-UHFFFAOYSA-N
SMILES:
COC1=CC=C(C=C1)C1=C(C=O)C=NN1
1
–
1
of
1
results
1
3-(4-Methoxyphenyl)-1H-pyrazole-4-carbaldehyde, 95%, Thermo Scientific™
CAS: 199682-73-0 Molecular Formula: C11H10N2O2 Molecular Weight (g/mol): 202.213 MDL Number: MFCD01133696 InChI Key: QSGGFCPKXTULQQ-UHFFFAOYSA-N Synonym: 3-4-methoxyphenyl-1h-pyrazole-4-carbaldehyde,5-4-methoxyphenyl-1h-pyrazole-4-carbaldehyde,3-4-methoxyphenyl-1h-pyrazole-4-carboxaldehyde,3-4-methoxyphenyl pyrazole-4-carbaldehyde,3-4-methoxyphenyl-2h-pyrazole-4-carbaldehyde,cbmicro_017678,cambridge id 6132430,3-4-methoxyphenyl pyrazole-4-carboxaldehyde,3-4-methoxy-phenyl-1h-pyrazole-4-carbaldehyde,1h-pyrazole-4-carbaldehyde, 3-4-methoxyphenyl PubChem CID: 737224 IUPAC Name: 5-(4-methoxyphenyl)-1H-pyrazole-4-carbaldehyde SMILES: COC1=CC=C(C=C1)C2=C(C=NN2)C=O