Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(4-Methoxyphenyl)-1H-pyrazole-4-carbaldehyde, 95%, Thermo Scientific™

Catalog No. SEW05757DA
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
SEW05757DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SEW05757DA Supplier Thermo Scientific Chemicals Supplier No. SEW05757DA
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 199682-73-0
Molecular Formula C11H10N2O2
Molecular Weight (g/mol) 202.213
MDL Number MFCD01133696
InChI Key QSGGFCPKXTULQQ-UHFFFAOYSA-N
Synonym 3-4-methoxyphenyl-1h-pyrazole-4-carbaldehyde, 5-4-methoxyphenyl-1h-pyrazole-4-carbaldehyde, 3-4-methoxyphenyl-1h-pyrazole-4-carboxaldehyde, 3-4-methoxyphenyl pyrazole-4-carbaldehyde, 3-4-methoxyphenyl-2h-pyrazole-4-carbaldehyde, cbmicro_017678, cambridge id 6132430, 3-4-methoxyphenyl pyrazole-4-carboxaldehyde, 3-4-methoxy-phenyl-1h-pyrazole-4-carbaldehyde, 1h-pyrazole-4-carbaldehyde, 3-4-methoxyphenyl
PubChem CID 737224
IUPAC Name 5-(4-methoxyphenyl)-1H-pyrazole-4-carbaldehyde
SMILES COC1=CC=C(C=C1)C2=C(C=NN2)C=O

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 202.21
Percent Purity 95%
Chemical Name or Material 3-(4-Methoxyphenyl)-1H-pyrazole-4-carbaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.