accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 19978-61-1

IUPAC Name: [2-(diphenylphosphanyl)ethyl]diphenylphosphane; dichloropalladium
Synonyms: diphos; palladium chloride
Molecular Weight (g/mol): 575.75
Molecular Formula: C26H24Cl2P2Pd
InChi Key: LDJXFZUGZASGIW-UHFFFAOYSA-L
SMILES: Cl[Pd]Cl.C(CP(C1=CC=CC=C1)C1=CC=CC=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)
  • (2)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)