Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (1R)-(-)-Nopol

Catalog No. 340367100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
34-036-7100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning
Catalog No. 34-036-7100ML Supplier Sigma Aldrich Supplier No. 340367100ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

(1R)-(-)-Nopol is a chiral terpenol derivative.(1R)-(-)-Nopol may be used in the preparation of optically active O,O-diterpenyl dithiophosphoric and O-terpenyl aryldithiophosphonic acids by reacting with tetraphosphorus decasulfide and 2,4-diaryl 1,3,2,4-dithiadiphosphetane-2,4-disulfides, respectively. It may also react with silver hexafluoroantimonate to form a complex, which can be an effective initiator for the nonthermal curing of epoxy resins by electron beam induced cationic polymerization.

Specifications

CAS 19978-61-1
Molecular Formula C26H24Cl2P2Pd
Refractive Index n20/D 1.493
Linear Formula [(C6H5)2PCH2CH2P(C6H5)2]PdCl2
MDL Number MFCD00015702
Quantity 100 mL
Synonym (-)-Nopol; (1R)-6,6-Dimethylbicyclo[3.1.1]hept-2-ene-2-ethanol; 6,6-Dimethyl-2-norpinene-2-ethanol; 6,6-Dimethylbicyclo[3.1.1]hept-2-ene-2-ethanol
Molecular Weight (g/mol) 575.74 g/mol
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.